missing translation for 'onlineSavingsMsg'
Learn More

Methyltestosterone (17alpha-Methyltestosterone), LoGiCal

Product Code. 30457649
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457649 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457649 Supplier LoGiCal Supplier No. LGCFOR0558.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 17-alpha-Methyltestosterone
CAS 58-18-4
Analyte or Component Names 17-alpha-Methyltestosterone
Shipping Condition Room Temperature
Molecular Formula C20 H30 O2
Certifications/Compliance ISO 9001
Synonym Methyltestosterone, 17alpha-Methyltestosterone, 17-Hydroxy-17-methyl-3-keto-androstene-4, 17-Methyltestosteron, 17-Methyltestosterone, Android, Andrometh, Androsan, Androsan (tablets), Androsten, Anertan, Anertan (tablets), CDB 110, Dumogran, Glosso-Sterandryl, Homandren, Hormale, L 589.372, M.T.Mucorettes, Malestrone, Malogen, Masenone, Mastestona, Mesterone, Metandren, Methyltestorone, Metrone, NSC 139965, NSC 9701, Neo-Hombreol-M, Nu-man, Oraviron, Orchisterone M, Oreton Methyl, Oreton-M, Perandren, RU 24400, Steronyl, Synandrets, Synandrotabs, Testhormone, Testora, Testoviron, Testoviron (tablet), Testovis Depot, Testred, U 2842, Androst-4-en-3-one, 17-hydroxy-17-methyl-, (17beta)-, Androst-4-en-3-one, 17beta-hydroxy-17-methyl- (8CI), (17beta)-17-Hydroxy-17-methylandrost-4-en-3-one, 17(alpha)-Methyl-Delta4-androsten-17-(beta)-ol-3-one, 17alpha-Methyl-17beta-hydroxyandrost-4-en-3-one, 17alpha-Methyl-17beta-hydroxyandrost-4-ene-3-one, 17alpha-Methyl-3-oxo-4-androsten-17beta-ol, 17alpha-Methylandrost-4-en-17-ol-3-one, 17alpha-Methylandrost-4-en-17beta-ol-3-one, 17beta-Hydroxy-17-methylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-methyl-3-oxoandrost-4-ene, 17beta-Hydroxy-17alpha-methylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-methylandrost-4-ene-3-one, Androst-4-ene-17alpha-methyl-17beta-ol-3-one
InChI Formula InChI=1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h12,15-17,22H,4-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1
Solution Type Neat
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13,17-trimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Industry Type Clinical Testing
Molecular Weight (g/mol) 302.45
Quantity 10 mg
Formula Weight 302.2246 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.