missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfuric Acid Potassium Salt, TRC

Product Code. 30134076
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134076 10 g 10g
30135371 5 g 5g
30008629 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134076 Supplier Toronto Research Chemicals Supplier No. TRCM33995510G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-54-9
Molecular Formula CH3O4SK
Molecular Weight (g/mol) 150.2
Synonym Methyl Potassium Sulfate;Potassium Methyl Sulfate;Potassium Monomethylsulfate
IUPAC Name potassium;methyl sulfate
SMILES [K+].COS(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/CH4O4S.K/c1-5-6(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1
Formula Weight 149.9389
Chemical Name or Material Methylsulfuric Acid Potassium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.