missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfuric acid potassium salt, 97%

Product Code. 10342333 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10342333 10 g Glass bottle 10g
10405661 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10342333 Supplier Thermo Scientific Acros Supplier No. 414890100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 562-54-9 | CH3KO4S | 150.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 562-54-9
Molecular Formula CH3KO4S
Molecular Weight (g/mol) 150.19
MDL Number MFCD00040472
InChI Key WBGWGHYJIFOATF-UHFFFAOYSA-M
Synonym potassium methyl sulfate, methyl potassium sulfate, methylsulfuric acid potassium salt, unii-9b0jf05o1c, sulfuric acid, monomethyl ester, potassium salt, potassium methyl sulphate, potassium monomethylsulfate, sulfuric acid, monomethyl ester, potassium salt 1:1, 75-93-4 parent, potassium methylsulfate
PubChem CID 2733094
IUPAC Name potassium;methyl sulfate
SMILES [K+].COS([O-])(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 212.0°C to 217.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3OSO3K
Quantity 10 g
Fieser 01,1249; 02,446; 04,557; 06,649; 08,354
Merck Index 13, 7731
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol
Formula Weight 150.19
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Methylsulfuric acid potassium salt, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.