missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfonylacetone, 98+%

Product Code. 11392146
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11392146 50 g 50g
11372146 2 g 2g
11382146 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11392146 Supplier Thermo Scientific Alfa Aesar Supplier No. L03421.18
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5000-46-4 | C4H8O3S | 136.165 g/mol

Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylsulfonylacetone is used as reactant for Stereoselective preparation of chiral cyclic ketones. It is used in the Preparation of poly-substituted pyridines. It aids for Gold-catalyzed Friedlander cyclocondensation reaction. It is used as reactant in Radical homoallylation reactions, multicomponent cyclocondensation with aldehydes and aminoazoles.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5000-46-4
Molecular Formula C4H8O3S
Molecular Weight (g/mol) 136.165
MDL Number MFCD00014745
InChI Key NWEYGXQKFVGUFR-UHFFFAOYSA-N
Synonym methylsulfonylacetone, 1-methylsulfonyl propan-2-one, methanesulfonylacetone, 1-methylsulfonyl acetone, 1-methanesulfonylpropan-2-one, 1-methylsulphonyl acetone, 2-propanone, 1-methylsulfonyl, methylsulfonylaceton, methylsulphonylacetone, methanesulphonylacetone
PubChem CID 78695
IUPAC Name 1-methylsulfonylpropan-2-one
SMILES CC(=O)CS(=O)(=O)C

Specifications

Melting Point 47°C to 52°C
Quantity 50 g
Beilstein 1754113
Solubility Information Insoluble in water.
Formula Weight 136.18
Percent Purity ≥98%
Chemical Name or Material Methylsulfonylacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.