missing translation for 'onlineSavingsMsg'
Learn More

Methylsuccinic Acid, TRC

Product Code. 17973645
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17973645 1 g 1g
17912405 10 g 10g
17951225 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17973645 Supplier Toronto Research Chemicals Supplier No. TRCM3290451G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-21-5
Molecular Formula C5 H8 O4
Molecular Weight (g/mol) 132.12
Synonym 2-Methylbutanedioic acid
IUPAC Name 2-methylbutanedioic acid
SMILES CC(CC(=O)O)C(=O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H8O4/c1-3(5(8)9)2-4(6)7/h3H,2H2,1H3,(H,6,7)(H,8,9)
Formula Weight 132.0423
Chemical Name or Material Methylsuccinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.