missing translation for 'onlineSavingsMsg'
Learn More

Methylpropyldichlorosilane, 97%, Thermo Scientific™

Product Code. 11995911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11995911 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11995911 Supplier Thermo Scientific Acros Supplier No. 432880250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Specifications

CAS 4518-94-9
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0300g/mL
Boiling Point 125.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H10Cl2Si
Quantity 25 mL
Specific Gravity 1.03
Synonym methylpropyldichlorosilane, silane, dichloromethylpropyl, dichloromethylpropylsilane, n-propylmethyldichlorosilane, dichloro methyl propylsilane, dichloromethylpropyl-silan, n-propyl methyl dichlorosilane, dichloro methyl propyl silane, dichloro methyl propylsilane gc
InChI Key GNVPGBIHGALKRR-UHFFFAOYSA-N
SMILES CCC[Si](C)(Cl)Cl
IUPAC Name dichloro-methyl-propylsilane
Molecular Weight (g/mol) 157.12
PubChem CID 78275
Formula Weight 157.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylpropyldichlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methylpropyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.