missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Methylproamine

Product Code. 16436846
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16436846 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16436846 Supplier MedChemExpress Supplier No. HY15620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methylproamine is a DNA-binding radioprotector, acts by repair of transient radiation-induced oxidative species on DNA. Methylproamine also protects against ionizing radiation by preventing DNA double-strand breaks.

Specifications

Molecular Weight (g/mol) 465.59 g/mol
Product Type Biochemical Assay Reagents
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Shipping Condition Room temperature
Purity 95.32%
Quantity 100 mg
Solubility Information DMSO : ≥ 41 mg/mL (88.06 mM)
For Use With (Application) Neuroscience-Neuromodulation
CAS 188247-01-0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.