missing translation for 'onlineSavingsMsg'
Learn More

Methylprednisolone, Mikromol™

Product Code. 18053990
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
18053990 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 18053990

Brand: Mikromol MM0190.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methylprednisolone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylprednisolone
CAS 83-43-2
Analyte or Component Names Methylprednisolone
Shipping Condition Room Temperature
Molecular Formula C22 H30 O5
Certifications/Compliance ISO 17034
Synonym Methylprednisolone, Methylprednisolone Hydrogen Succinate Imp. A (EP), Methylprednisolone Acetate Imp. B (EP), 6α-methylprednisolone, Methylprednisolone Acetate Impurity B, Methylprednisolone Hydrogen Succinate Impurity A, Methylprednisolone Hydrogen Succinate Impurity A
InChI Formula InChI=1S/C22H30O5/c1-12-8-14-15-5-7-22(27,18(26)11-23)21(15,3)10-17(25)19(14)20(2)6-4-13(24)9-16(12)20/h4,6,9,12,14-15,17,19,23,25,27H,5,7-8,10-11H2,1-3H3/t12-,14-,15-,17-,19+,20-,21-,22-/m0/s1
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)CO)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
IUPAC Name (6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-6,10,13-trimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 374.47
Quantity 250 mg
Formula Weight 374.2093 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.