missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylprednisolone Hydrogen Succinate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Methylprednisolone Hydrogen Succinate
Specifications
Specifications
| Chemical Name or Material | Methylprednisolone Hydrogen Succinate |
| CAS | 2921-57-5 |
| Analyte or Component Names | Methylprednisolone Hydrogen Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H34 O8 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Methylprednisolone hydrogen succinate, 6-Methylprednisolone hemisuccinate, Methylprednisolone 21-hemisuccinate, Methylprednisolone hemisuccinate, Methylprednisolone succinate, 4-[(11beta,17-Dihydroxy-6alpha-methyl-3,20-dioxopregna-1,4-dien-21-yl)oxy]-4-oxobutanoic acid, (6alpha,11beta)-21-(3-carboxy-1-oxopropoxy)-11,17-dihydroxy-6-methyl-pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-6alpha-methyl-, 21-(H succinate) (7CI), Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-6alpha-methyl-, 21-(hydrogen succinate) (8CI), Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-6alpha-methyl-, 21-succinate (6CI), 6alpha-Methylprednisolone 21-succinate, 6alpha-Methylprednisolone hemisuccinate |
| InChI Formula | InChI=1S/C26H34O8/c1-14-10-16-17-7-9-26(33,20(29)13-34-22(32)5-4-21(30)31)25(17,3)12-19(28)23(16)24(2)8-6-15(27)11-18(14)24/h6,8,11,14,16-17,19,23,28,33H,4-5,7,9-10,12-13H2,1-3H3,(H,30,31)/t14-,16-,17-,19-,23+,24-,25-,26-/m0/s1 |
| SMILES | C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)CCC(=O)O)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14 |
| IUPAC Name | 4-[2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]-4-oxobutanoic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction