missing translation for 'onlineSavingsMsg'
Learn More

Methylprednisolone Acetate, LoGiCal

Product Code. 30461135
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461135 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461135 Supplier LoGiCal Supplier No. LGCFOR0191.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methylprednisolone Acetate
CAS 53-36-1
Analyte or Component Names Methylprednisolone Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H32 O6
Certifications/Compliance ISO 9001
Synonym Methylprednisolone Acetate, Methylprednisolone Hydrogen Succinate Imp. C (EP), 6-Methylprednisolone acetate, DepMedalone 40, DepMedalone 80, Depo-Medrate, Depo-Medrol, Depo-Medrone, Depo-methylprednisolone, MPA, Medrol acetate, Mepred, Methylprednisolone 21-acetate, Methylprednisolone acetate, NSC 48985, U 8210, Urbason Crystal Suspension, Vetacortyl, Pregna-1,4-diene-3,20-dione, 11beta,17,21-trihydroxy-6alpha-methyl-, 21-acetate (6CI,7CI,8CI), (6alpha,11beta)-21-(Acetyloxy)-11,17-dihydroxy-6-methylpregna-1,4-diene-3,20-dione, 11beta,17alpha,21-Trihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione 21-acetate, 21-Acetoxy-11beta,17-dihydroxy-6alpha-methylpregna-1,4-diene-3,20-dione, 6alpha-Methylprednisolone 21-acetate, 6alpha-Methylprednisolone acetate, Methylprednisolone Hydrogen Succinate Impurity C, Methylprednisolone Impurity J
InChI Formula InChI=1S/C24H32O6/c1-13-9-16-17-6-8-24(29,20(28)12-30-14(2)25)23(17,4)11-19(27)21(16)22(3)7-5-15(26)10-18(13)22/h5,7,10,13,16-17,19,21,27,29H,6,8-9,11-12H2,1-4H3/t13-,16-,17-,19-,21+,22-,23-,24-/m0/s1
Solution Type Neat
SMILES C[C@H]1C[C@H]2[C@@H]3CC[C@](O)(C(=O)COC(=O)C)[C@@]3(C)C[C@H](O)[C@@H]2[C@@]4(C)C=CC(=O)C=C14
IUPAC Name [2-[(6S,8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-6,10,13-trimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 416.51
Quantity 10 mg
Formula Weight 416.2199 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.