missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Methylphenylphosphinic chloride, 97%

Product Code. 11403917
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11403917

Brand: Alfa Aesar™ A18349.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5761-97-7
Molecular Formula C7H8ClOP
Molecular Weight (g/mol) 174.56
MDL Number MFCD00013902
InChI Key BTMQENOUJJKBGT-UHFFFAOYNA-N
Synonym methylphenylphosphinic chloride, chloro methyl phosphoryl benzene, methyl phenyl phosphinoyl chloride, acmc-1arvk, methyl phenylphosphinicchloride, methylphenylphosphinyl chloride, methylphenyl-phosphinic chloride, phenyl methyl phosphinyl chloride, phosphinic chloride,methylphenyl
PubChem CID 21983
IUPAC Name [chloro(methyl)phosphoryl]benzene
SMILES CP(Cl)(=O)C1=CC=CC=C1

Specifications

Density 1.266
Boiling Point 155°C (11 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 174.57
Percent Purity 97%
Chemical Name or Material Methylphenylphosphinic chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.