missing translation for 'onlineSavingsMsg'
Learn More

Methylmercury(II) chloride, standard solution in H{2}O, Methylmercury(II) chloride 1000ppm by AA

Product Code. 11377758
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377758 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377758 Supplier Thermo Scientific Alfa Aesar Supplier No. 033553.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 115-09-3 | CH3ClHg | 251.077 g/mol

Methylmercury(II) chloride is used as a precursor for the preparation of methyl mercury acetate. It is used as a specific reagent (electron stain) for sulfhydryl groups in biological materials like protein as well as demonstration in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-09-3
Molecular Formula CH3ClHg
Molecular Weight (g/mol) 251.077
MDL Number MFCD00013592
InChI Key BABMCXWQNSQAOC-UHFFFAOYSA-M
Synonym methylmercury chloride, methylmercuric chloride, caspan, mercury, chloromethyl, methyl mercuric chloride, mercury methyl chloride, chloro methyl mercury, methyl meruric chloride, monomethyl mercury chloride, methylmercury monochloride
PubChem CID 409301
ChEBI CHEBI:28216
IUPAC Name chloro(methyl)mercury
SMILES C[Hg]Cl

Specifications

Physical Form Liquid
Quantity 5 mL
Beilstein 3600218
Solubility Information Miscible with lipid.
Formula Weight 251.08
Appearance Clear
Chemical Name or Material Methylmercury(II) chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.