missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonyl Coenzyme A mutase Antibody (OTI2C8), Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18416255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416255 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416255 Supplier Novus Biologicals Supplier No. NBP272652AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Methylmalonyl Coenzyme A mutase Monoclonal antibody specifically detects Methylmalonyl Coenzyme A mutase in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Methylmalonyl Coenzyme A mutase
Applications Immunofluorescence
Classification Monoclonal
Clone OTI2C8
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias EC 5.4.99.2, MCMmethylmalonyl Coenzyme A mutase, methylmalonyl CoA mutase, Methylmalonyl-CoA isomerase, methylmalonyl-CoA mutase, mitochondrial
Host Species Mouse
Immunogen Full length recombinant protein of human Methylmalonyl Coenzyme A mutase (NP_000246) produced in HEK293T cells.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4594
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.