missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonic Acid, TRC

Product Code. 30007322
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007322 5 g 5g
30003582 1 g 1g
30005693 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007322 Supplier Toronto Research Chemicals Supplier No. TRCM3188605G
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-05-2
Molecular Formula C4 H6 O4
Molecular Weight (g/mol) 118.09
Synonym Malonic acid, methyl- (6CI,8CI), Propanedioic acid, methyl- (9CI), 2-Methylpropanedioic acid, 1,1-Ethanedicarboxylic acid, 2-Methylmalonic acid, Isosuccinic acid, Methylmalonic acid, Methylpropanedioic acid, NSC 25201
IUPAC Name 2-methylpropanedioic acid
SMILES CC(C(=O)O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H6O4/c1-2(3(5)6)4(7)8/h2H,1H3,(H,5,6)(H,7,8)
Formula Weight 118.0266
Percent Purity >95%
Chemical Name or Material Methylmalonic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.