missing translation for 'onlineSavingsMsg'
Learn More

Methylmalonic acid, 96%

Product Code. 10605372 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10605372 25 g Glass bottle 25g
10583671 5 g Glass bottle 5g
10580774 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10605372 Supplier Thermo Scientific Acros Supplier No. 127090250
  • ONLINE EXCLUSIVE PRICE
    £119.00
    £107.65 / 25g
    Save £11.35
    9% Off
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 516-05-2 | C4H6O4 | 118.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 516-05-2
Molecular Formula C4H6O4
Molecular Weight (g/mol) 118.09
MDL Number MFCD00002656
InChI Key ZIYVHBGGAOATLY-UHFFFAOYSA-N
Synonym methylmalonic acid, 2-methylmalonic acid, isosuccinic acid, methylmalonate, propanedioic acid, methyl, 1,1-ethanedicarboxylic acid, malonic acid, methyl, methyl malonic acid, methylpropanedioic acid, unii-8ll8s712j7
PubChem CID 487
ChEBI CHEBI:30860
IUPAC Name 2-methylpropanedioic acid
SMILES CC(C(O)=O)C(O)=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 129.0°C to 136.0°C
Color White
Density 1.4500g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula HO2CCH(CH3)CO2H
Quantity 25 g
Beilstein 02, IV, 1932
Specific Gravity 1.45
Solubility Information Solubility in water: very soluble. Other solubilities: very soluble in ethanol,soluble in ether and ethyl acetate,slightly soluble in benzene and trifluoroacetic,acid
Formula Weight 118.09
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Methylmalonic acid
Show More Show Less
compliance-icons
Product Identifier
  • Methylmalonic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.