missing translation for 'onlineSavingsMsg'
Learn More

Methylmagnesium iodide, 3M solution in diethyl ether, AcroSeal™

Product Code. 10632445 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
800mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10632445 800 mL AcroSeal™ Glass Bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10632445 Supplier Thermo Scientific Acros Supplier No. 427408000
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 917-64-6 | CH3IMg | 166.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-64-6 , 60-29-7
Molecular Formula CH3IMg
Molecular Weight (g/mol) 166.24
MDL Number MFCD00001026
InChI Key AUPXBVDHVRZMIB-UHFFFAOYSA-M
Synonym methylmagnesium iodide, magnesium, iodomethyl, methylmagnesiumiodide, methyl magnesium iodide, iodomethylmagnesium, methylmagnesium iodide solution, 3.0 m in diethyl ether, memgi, ch3mgi, methyl-magnesium iodide
PubChem CID 101907
IUPAC Name magnesium;carbanide;iodide
SMILES C[Mg]I

Specifications

Density 1.2600g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3MgI
Quantity 800 mL
Specific Gravity 1.26
Formula Weight 166.24
Chemical Name or Material Methylmagnesium iodide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylmagnesium iodide, 3M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which, in contact with water, emits flammable gases Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P233-Keep container tightly closed.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.