missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyllycaconitine citrate, Tocris Bioscience™
Selective α7 neuronal nAChR antagonist
£106.24 - £358.52
Chemical Identifiers
CAS | 351344-10-0 |
---|---|
Molecular Formula | C43H58N2O17 |
Molecular Weight (g/mol) | 874.934 |
InChI Key | INBLZNJHDLEWPS-OXVGBIBJSA-N |
Synonym | methyllycaconitine citrate |
PubChem CID | 124081005 |
SMILES | CCN1CC2(CCC(C34C2C(C(C31)(C5(CC(C6CC4C5C6OC)OC)O)O)OC)OC)COC(=O)C7=CC=CC=C7N8C(=O)CC(C8=O)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18074262
|
Tocris Bioscience
1029/5 |
5 mg |
£106.24
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30001166
|
Tocris Bioscience
1029/25 |
25 mg |
£358.52
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent antagonist for α7-containing neuronal nicotinic receptors (Ki = 1.4 nM). Interacts with α4β2 and α6β2 receptors at concentrations > 40 nM. Attenuates METH-induced neurotoxicity in mouse striatum in vivo.Chemical Identifiers
351344-10-0 | |
874.934 | |
methyllycaconitine citrate | |
CCN1CC2(CCC(C34C2C(C(C31)(C5(CC(C6CC4C5C6OC)OC)O)O)OC)OC)COC(=O)C7=CC=CC=C7N8C(=O)CC(C8=O)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O |
C43H58N2O17 | |
INBLZNJHDLEWPS-OXVGBIBJSA-N | |
124081005 |
Specifications
351344-10-0 | |
methyllycaconitine citrate | |
CCN1CC2(CCC(C34C2C(C(C31)(C5(CC(C6CC4C5C6OC)OC)O)O)OC)OC)COC(=O)C7=CC=CC=C7N8C(=O)CC(C8=O)C.C(C(=O)O)C(CC(=O)O)(C(=O)O)O | |
124081005 | |
95% | |
Methyllycaconitine citrate |
C43H58N2O17 | |
INBLZNJHDLEWPS-OXVGBIBJSA-N | |
874.934 | |
874.93 | |
Solid |
Safety and Handling
Recommended Storage : Store at -20°C