missing translation for 'onlineSavingsMsg'
Learn More

Methyllycaconitine citrate, MedChemExpress

Product Code. 16491708
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16491708

Brand: Medchem Express HYN2332A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Methyllycaconitine citrate is a specific antagonist of α7 neuronal nicotinic acetylcholine receptor (α7nAChR).

Methyllycaconitine citrate is a specific antagonist of α7 neuronal nicotinic acetylcholine receptor (α7nAChR).

Specifications

Chemical Name or Material Methyllycaconitine citrate
CAS 351344-10-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C43H58N2O17
Quantity 5 mg
Synonym MLA
Solubility Information DMSO : 125 mg/mL (142.87 mM; Need ultrasonic) ∣H2O : 1.67 mg/mL (1.91 mM; Need ultrasonic)
SMILES CO[C@@H]1C(C(N(CC)C2)C3(O)[C@H]4OC)([C@@](C[C@@]5([H])[C@@H]6OC)([H])[C@@]6([H])[C@@]3(C[C@@H]5OC)O)[C@@]4([H])[C@@]2(COC(C7=C(N(C8=O)C(C[C@@H]8C)=O)C=CC=C7)=O)CC1.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 874.92
Formula Weight 874.92
Percent Purity 97.24%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.