missing translation for 'onlineSavingsMsg'
Learn More

Methyllithium, 2.2M (6wt%) in diethyl ether with LiBr, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10547573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10547573 800 mL AcroSeal™ Glass Bottle 800mL
10547383 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10547573 Supplier Thermo Scientific Acros Supplier No. 181298000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 917-54-4 | Molecular Formula: CH3Li | Molecular Weight: 21.98 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-54-4 , 60-29-7
Molecular Formula CH3Li
Molecular Weight (g/mol) 21.98
MDL Number MFCD00008253
InChI Key DVSDBMFJEQPWNO-UHFFFAOYSA-N
Synonym methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
PubChem CID 2724049
IUPAC Name lithium;carbanide
SMILES [Li]C

Specifications

Color Colorless through Yellow
Physical Form Solution
Quantity 800 mL
Linear Formula CH3Li
Flash Point −28°C
Solubility Information Solubility in water: vigorous reaction
Formula Weight 21.98
Percent Purity 97%
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.81
Density 0.8100g/mL
Chemical Name or Material Methyllithium
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.