missing translation for 'onlineSavingsMsg'
Learn More

Methyllithium, 2.2M (6wt%) in diethyl ether with LiBr, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10547383 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10547383 100 mL AcroSeal™ Glass bottle 100mL
10547573 800 mL AcroSeal™ Glass bottle 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10547383 Supplier Thermo Scientific Acros Supplier No. 181291000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-54-4 , 60-29-7
Molecular Formula CH3Li
Molecular Weight (g/mol) 21.98
MDL Number MFCD00008253
InChI Key DVSDBMFJEQPWNO-UHFFFAOYSA-N
Synonym methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
PubChem CID 2724049
IUPAC Name lithium;carbanide
SMILES [Li]C

Specifications

Color Colorless through Yellow
Physical Form Solution
Quantity 100 mL
Linear Formula CH3Li
Flash Point −28°C
Solubility Information Solubility in water: vigorous reaction
Formula Weight 21.98
Percent Purity 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.81
Density 0.8100g/mL
Chemical Name or Material Methyllithium
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methyllithium, 2.2M (6wt%) in diethyl ether with lithiumbromide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 1
  • Pyrophoric solid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Skin sensitiser Category 1
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H336-May cause drowsiness or dizziness.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Lithium bromide (LiBr), Lithium, methyl-, Ethyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.