missing translation for 'onlineSavingsMsg'
Learn More

Methyliminodiacetic Acid, MP Biomedicals

Product Code. 11441116 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
5g

Product Code. 11441116

Brand: MP Biomedicals 0520248205

Please to purchase this item. Need a web account? Register with us today!

Specifications

CAS 4408-64-4
Molecular Formula C5H9NO4
Quantity 10 g
Synonym n-methyliminodiacetic acid, 2,2'-methylazanediyl diacetic acid, methyliminodiacetic acid, mida, methylimidodiacetic acid, n-methyliminodiaceticacid, glycine, n-carboxymethyl-n-methyl, methylimino diacetic acid, acetic acid, methylimino di, carboxymethyl methyl amino acetic acid
InChI Key XWSGEVNYFYKXCP-UHFFFAOYSA-N
SMILES CN(CC(=O)O)CC(=O)O
IUPAC Name 2-[carboxymethyl(methyl)amino]acetic acid
Molecular Weight (g/mol) 147.13
PubChem CID 20441
Formula Weight 147.13
Chemical Name or Material Methyliminodiacetic Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.