missing translation for 'onlineSavingsMsg'
Learn More

alpha-Methylhydrocinnamic acid, 98%

Product Code. 10016222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10016222 5 g Glass bottle 5g
10684853 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10016222 Supplier Thermo Scientific Acros Supplier No. 379080050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1009-67-2 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1009-67-2
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00192301
InChI Key MCIIDRLDHRQKPH-UHFFFAOYNA-N
Synonym 2-benzylpropionic acid, alpha-methylhydrocinnamic acid, 2-methyl-3-phenylpropionic acid, 2-benzylpropanoic acid, 2-methyl-3-phenyl-propionic acid, acmc-20ao3h, alpha-methylhydrocinnamicacid, alpha-methyl-hydrocinnamic acid, 2-methyl-3-phenyl-propanoic acid, #
PubChem CID 99862
IUPAC Name 2-methyl-3-phenylpropanoic acid
SMILES CC(CC1=CC=CC=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 37°C to 41°C
Color White to Yellow
Boiling Point 167°C to 168°C (23.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH2CH(CH3)CO2H
Quantity 5 g
Formula Weight 164.2
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material α-Methylhydrocinnamic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.