missing translation for 'onlineSavingsMsg'
Learn More

Methylhydrazine sulfate, 98%

Product Code. p-7045278
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11378235

Brand: Thermo Scientific Alfa Aesar L10122.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 302-15-8 | CH8N2O4S | 144.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-15-8
Molecular Formula CH8N2O4S
Molecular Weight (g/mol) 144.15
MDL Number MFCD00035423
InChI Key KJDJPXUIZYHXEZ-UHFFFAOYSA-N
Synonym methylhydrazine sulfate, methylhydrazine; sulfuric acid, methyl hydrazine sulfate, methyl hydrazine sulphate, hydrazine, methyl-, sulfate, methyl hydrazine; sulfuric acid, ch6n2.h2o4s, methylhydrazine sulphate, dsstox_cid_875, n-methyl hydrazine sulfate
PubChem CID 165286
IUPAC Name methylhydrazine;sulfuric acid
SMILES CN[NH3+].OS([O-])(=O)=O

Specifications

Melting Point 141°C to 143°C
Quantity 5 g
UN Number UN2811
Beilstein 3693575
Merck Index 14,6086
Formula Weight 144.15
Percent Purity 98%
Chemical Name or Material Methylhydrazine sulfate
compliance-icons compliance-icons
Product Identifier
  • Methylhydrazine sulphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H330-Fatal if inhaled.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methylhydrazine sulphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.