missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methylglyoxal Monoclonal Antibody (9F11), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA545816
This item is not returnable.
View return policy
Description
A 1:1,000 dilution of MA5-45816 was sufficient for detection of Methylglyoxal in 0.5 μg of Methylglyoxal conjugated to BSA by ECL immunoblot analysis using Goat Anti-Mouse IgG:HRP as the secondary Antibody.∣Specific for Methylglyoxal modified proteins. Does not detect free Methylglyoxal. Does not cross-react with Acrolein, Hexanoyl Lysine, Malondialdeyhde, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, or Crotonaldehyde modified proteins.
Methylglyoxal, also called pyruvaldehyde or 2-oxopropanal, is the organic compound with the formula CH3C(O)CHO. Gaseous methylglyoxal has two carbonyl groups, an aldehyde and a ketone but in the presence of water, it exists as hydrates and oligomers. Methylglyoxal is a reduced derivative of pyruvic acid.Specifications
Methylglyoxal | |
Monoclonal | |
1 mg/mL | |
2.15mM MES, 47.35mM phosphate with 1.033mM EDTA and no preservative; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
Chemical | |
Liquid | |
Chemical |
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot | |
9F11 | |
PE | |
2 oxo propanal; 2-Oxopropanal; Aldoketomutase; epididymis secretory protein Li 74; GLO1; GLOD1; Glx I; GLYI; glyoxalase domain containing 1; glyoxalase I; HEL-S-74; ketone-aldehyde mutase; lactoyl glutathione lyase; lactoylglutathione lyase; Methylglyoxal; Methylglyoxal (MG); Methylglyoxalase; MG; MG-modified protein; Pyruvaldehyde; S-D-lactoylglutathione methylglyoxal lyase | |
Synthetic Methylglyoxal modified Keyhole Limpet Hemocyanin (KLH). | |
100 μg | |
Primary | |
4° C | |
IgG1 |