missing translation for 'onlineSavingsMsg'
Learn More

Methylgermanium trichloride, 97%

Product Code. 11384168
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384168 2 g 2g
11394168 0.5 g 0.5g
11304178 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384168 Supplier Thermo Scientific Alfa Aesar Supplier No. 041449.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 993-10-2 | CH9Cl3Ge | 200.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylgermanium trichloride is used as a precursor for the synthesis of five-coordinated spirocyclic anionic methylgermanates by reacting with catechol. It is also involved in the photo-chemical adsorption of organo-germanium on a hydrogen terminated atomically flat silicon surface using xenon excimer light.

Solubility
Miscible with ether.

Notes
Sensitive to moisture and reacts violently with water. Also hydrolysis takes place when contact with water. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 993-10-2
Molecular Formula CH9Cl3Ge
Molecular Weight (g/mol) 200.06
MDL Number MFCD00013585
InChI Key NNMJXXWNUALEKD-UHFFFAOYSA-N
Synonym methyltrichlorogermane, trichloro methyl germane, methylgermanium trichloride, germanium methyl trichloride, germane, trichloromethyl, fefxfmqvsdtspa-uhfffaoysa, inchi=1/ch3cl3ge/c1-5 2,3 4/h1h3
PubChem CID 70436
IUPAC Name trichloro(methyl)germane
SMILES Cl.Cl.Cl.C[GeH3]

Specifications

Color Colorless to Yellow
Density 1.706 g/mL
Boiling Point 111°C
Flash Point 10°C (50°F)
Odor Chlorine-like
Linear Formula CH3GeCl3
Refractive Index 1.4685
Quantity 2 g
UN Number UN2924
Sensitivity Moisture sensitive
Formula Weight 193.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methylgermanium trichloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.