missing translation for 'onlineSavingsMsg'
Learn More

Methyl trimethylacetate, 99%

Product Code. 10071330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
Packaging:
Glass bottle
Unit Size:
100mL
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10071330 250 mL Glass bottle 250mL
10198083 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071330 Supplier Thermo Scientific Acros Supplier No. 127952500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-98-1 | C6H12O2 | 116.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-98-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00008843
InChI Key CNMFHDIDIMZHKY-UHFFFAOYSA-N
Synonym methyl pivalate, methyl trimethylacetate, propanoic acid, 2,2-dimethyl-, methyl ester, pivalic acid, methyl ester, unii-bfx9w386ox, bfx9w386ox, pivalic acid methyl ester, methyl pivaloate, methyltrimethylacetate, tert-c4h9cooch3
PubChem CID 69027
IUPAC Name methyl 2,2-dimethylpropanoate
SMILES CC(C)(C)C(=O)OC

Specifications

Melting Point -70°C
Color Colorless
Density 0.8700g/mL
Boiling Point 101°C
Flash Point <1°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)3CCO2CH3
Refractive Index 1.3890 to 1.391
Quantity 250 mL
Beilstein 02, 320
Specific Gravity 0.87
Viscosity 30 mPa.s (25°C)
Formula Weight 116.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl trimethylacetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl trimethylacetate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.