missing translation for 'onlineSavingsMsg'
Learn More

Methyl succinyl chloride, 97%

Product Code. 10143194 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143194 25 g Glass bottle 25g
10396824 5 g Glass bottle 5g
10644365 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10143194 Supplier Thermo Scientific Acros Supplier No. 108400250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1490-25-1 | C5H7ClO3 | 150.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1490-25-1
Molecular Formula C5H7ClO3
Molecular Weight (g/mol) 150.56
MDL Number MFCD00000750
InChI Key SRXOJMOGPYFZKC-UHFFFAOYSA-N
Synonym methyl 4-chloro-4-oxobutyrate, methyl 3-chloroformyl propionate, 3-carbomethoxypropionyl chloride, methyl 4-chloro-4-oxo-butanoate, 3-carbomethoxy propionyl chloride, butanoic acid, 4-chloro-4-oxo-, methyl ester, methoxycarbonylpropionyl chloride, 4-chloro-4-oxo-butanoicacimethylester, succinic acid monomethyl ester chloride, 4-chloro-4-oxobutyric acid methyl ester
PubChem CID 73888
IUPAC Name methyl 4-chloro-4-oxobutanoate
SMILES COC(=O)CCC(=O)Cl

Specifications

Density 1.2320g/mL
Boiling Point 58°C to 65°C (3.0 mmHg)
Flash Point 73°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3O2CCH2CH2COCl
Refractive Index 1.4402
Quantity 25 g
Beilstein 02, II, 553
Specific Gravity 1.232
Formula Weight 150.56
Percent Purity 97%
Chemical Name or Material Methyl succinyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • Methyl succinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.