missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyruvate, 94%, Thermo Scientific Chemicals

Catalog Number p-3779193
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
600-22-6
C4H6O3
102.09
MFCD00008754
CWKLZLBVOJRSOM-UHFFFAOYSA-N
methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
11748
CHEBI:51850
methyl 2-oxopropanoate
CC(=O)C(=O)OC
This item is not returnable. View return policy
600-22-6
C4H6O3
102.09
MFCD00008754
CWKLZLBVOJRSOM-UHFFFAOYSA-N
methyl pyruvate, pyruvic acid, methyl ester, pyruvic acid methyl ester, methyl 2-oxopropionate, propanoic acid, 2-oxo-, methyl ester, methyl-pyruvate, methylglyoxylic acid methyl ester, unii-3kjm65g5xl, 3kjm65g5xl, 2-oxo-propionic acid methyl ester
11748
CHEBI:51850
methyl 2-oxopropanoate
CC(=O)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.1300g/mL
Boiling Point 134.0°C to 137.0°C
Flash Point 39°C
Infrared Spectrum Authentic
Assay Percent Range 94%
Packaging Glass bottle
Linear Formula CH3COCO2CH3
Refractive Index 1.4065
Quantity 25 g
Beilstein 03, 616
Merck Index 15, 8135
Specific Gravity 1.13
Solubility Information Solubility in water: soluble.
Formula Weight 102.09
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Methyl pyruvate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'