missing translation for 'onlineSavingsMsg'
Learn More

Methylenediphosphonic acid, 98+%

Product Code. p-7022168
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11442547 1 g 1g
11452547 5 g 5g
11462547 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442547 Supplier Thermo Scientific Alfa Aesar Supplier No. A14803.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methylenediphosphonic acid, >-99%, CH6O6P2, CAS Number-1984-15-2, acido medronico, methylenediphosphonic acid, acide medronique, acidum medronicum, phosphonic acid, methylenebis, methylenebis phosphonic acid, methylene diphosphonate, mdp-bracco, methanediphosphonic acid, medronic acid, 25g, 1708494 | CAS: 1984-15-2 | CH4Na2O6P2 | 219.96 g/mol

Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylenediphosphonic acid is used as a radioactive imaging agent, which is primarily used in skeletal scintigraphy. Due to its absorption by a variety of tumors, it is useful for the detection of neoplasms.

Solubility
Soluble in water, dimethylsulfoxide and methanol. Slightly soluble in anhydrous ethanol. Insoluble in methylene chloride.

Notes
Store in a cool place. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1984-15-2
Molecular Formula CH4Na2O6P2
Molecular Weight (g/mol) 219.96
MDL Number MFCD00002139
InChI Key RXSYPQBNTPQOTQ-UHFFFAOYSA-L
Synonym methylenediphosphonic acid, medronic acid, acido medronico, methylene diphosphonate, acide medronique, acidum medronicum, phosphonic acid, methylenebis, methylenebis phosphonic acid, mdp-bracco, methanediphosphonic acid
PubChem CID 16124
ChEBI CHEBI:43945
IUPAC Name phosphonomethylphosphonic acid
SMILES [Na+].[Na+].OP([O-])(=O)CP(O)([O-])=O

Specifications

Melting Point 202°C to 204°C
Odor Odorless
Quantity 1 g
UN Number UN3261
Beilstein 1708494
Merck Index 14,5792
Formula Weight 176
Percent Purity ≥99%
Chemical Name or Material Methylenediphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.