missing translation for 'onlineSavingsMsg'
Learn More

Methylenebis(thioacetic acid), 97+%, Thermo Scientific Chemicals

Product Code. 11403677
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403677 5 g 5g
11413677 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403677 Supplier Thermo Scientific Alfa Aesar Supplier No. A18154.06
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2068-24-8
Molecular Formula C5H8O4S2
Molecular Weight (g/mol) 196.235
MDL Number MFCD00021766
InChI Key CZEVAMMETWVBOD-UHFFFAOYSA-N
Synonym methylenedithiodiacetic acid, methylenebisthioglycolic acid, acetic acid, 2,2'-methylenebis thio bis, acetic acid, methylenedithio di, 2,2'-methylenebis thio diacetic acid, methylenebis-thioglycollic acid, 2,2'-methylenebis sulfanediyl diacetic acid, 2-carboxymethyl thio methyl thio acetic acid, carboxymethyl sulfanyl methyl sulfanyl acetic acid
PubChem CID 74949
IUPAC Name 2-(carboxymethylsulfanylmethylsulfanyl)acetic acid
SMILES C(C(=O)O)SCSCC(=O)O

Specifications

Melting Point 127°C to 129°C
Quantity 5 g
Formula Weight 196.24
Percent Purity ≥97%
Chemical Name or Material Methylenebis(thioacetic acid)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.