missing translation for 'onlineSavingsMsg'
Learn More

Methylene green, MP Biomedicals™

Product Code. 11427992 Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11427992

Brand: MP Biomedicals 0215264405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2679-01-8
Molecular Formula C16H17ClN4O2S
Molecular Weight (g/mol) 364.85
InChI Key YYGBVRCTHASBKD-UHFFFAOYSA-M
Synonym C.I. 52020, 3, 7-Bis(Dimethylamino)-4-Nitro-Phenothiazin-5-Ium Chloride
ChEBI CHEBI:87675

Specifications

SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C(=C3S2)[N+](=O)[O-].[Cl-]
IUPAC Name [7-(dimethylamino)-4-nitrophenothiazin-3-ylidene]-dimethylazanium;chloride
PubChem CID 75889
Formula Weight 364.86
Color Black
Quantity 5 g
Physical Form Powder
Chemical Name or Material Methylene green
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.