missing translation for 'onlineSavingsMsg'
Learn More

Methylene chloride ≥ 99.8% stabilized, ChromAR™ for HPLC, Macron Fine Chemicals™

Product Code. 15538724
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15538724 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15538724 Supplier Macron Fine Chemicals Supplier No. 671425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For High Performance Liquid Chromatography

  • Ultraviolet Absorbance (1.00-cm cell vs. water):
    • 235 nm ≤ 0.5
    • 240 nm ≤ 0.2
    • 250 nm ≤ 0.07
    • 260 nm ≤ 0.01
    • UV Cut-off ≤ 229 nm

Specifications

CAS 75-09-2
Quantity 2.5 L
Residue after Evaporation ≤ 0.0005%
Assay Percent Range ≥ 99.8%
Titratable Acid ≤ 0.0005 meq/g
Molecular Formula CH2Cl2
MDL Number MFCD00000881
UN Number 1593
Water ≤ 0.02%
Synonym Dichloromethane
Boiling Point 39.8°C (1013 hPa)
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
SMILES ClCCl
IUPAC Name dichloromethane
Molecular Weight (g/mol) 84.93
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P281-Use personal protective equipment as required.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.