missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Trihydrate, Zinc Free, MP Biomedicals

Product Code. 11436451 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436451 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11436451 Supplier MP Biomedicals Supplier No. 0219399805
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An ethidium bromide alternative for the visualization of nucleic acids in gels.

Specifications

CAS 61-73-4
Molecular Formula C16H18ClN3S
Synonym Methylthionine chloride, C.I. 52015
Color Dark Blue
Quantity 5 g
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES CN(C)C1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
Molecular Weight (g/mol) 319.851
PubChem CID 6099
ChEBI CHEBI:6872
Formula Weight 373.9
Grade Not Available
Chemical Name or Material Methylene Blue, An ethidium bromide alternative for the visualization of nucleic acids in gels
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.