missing translation for 'onlineSavingsMsg'
Learn More

Pro-Lab Diagnostics™ Methylene Blue Stain

Product Code. 12881813
Click to view available options
Quantity:
500mL
Unit Size:
500mL
This item is not returnable. View return policy

Product Code. 12881813

Brand: Pro-Lab Diagnostics™ PL.7027

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use as a counterstain during Ziehl-Neelsen and Kinyoun staining. Pro-Lab Diagnostics™ Methylene Blue Stain is a ready-to-use stain to differentiate non-acid-fast organisms from mycobacterium. It is manufactured to the highest standards, according to all regulatory requirements, and CE labelled.

Pro-Lab Diagnostics™ Microbiological Stain solutions are manufactured in purpose-designed laboratories to the highest standards using the finest raw materials. Strict quality control and filtering techniques are employed prior to release to ensure that all solutions offer excellent performance characteristics.

Acid-fast organisms, such as Mycobacterium, have wax-like, nearly impermeable cell walls that make them highly resistant to typical microbiological stains. They require staining via either the Ziehl-Neelsen (ZN) method, which requires heating, or the Kinyoun method, which does not. The basic method seen in Gram staining is used: a primary stain is applied, a differentiator removes the stain from non-acid-fast organisms and a counterstain is then applied to identify the decolored organisms.

  • Ready-to-use solution in a convenient, economical size
  • Acts as a counter-stain for non-acid-fast organisms in the Ziehl-Neelsen or Kinyoun staining method
  • Apply only after staining with Carbol Fuchsin and using a differentiator/decolorizer
  • Decolorized non-acid-fast cells take up the contrasting blue color for easy detection

Specifications

Solution Type Reagent
Type Methylene Blue
Quantity 500mL
Certifications/Compliance (EC) No 1907/2006, Annex II, (EU) No 453/2010, CE-mark
Format Liquid
For Use With (Application) For Mycobacteria acid fast stains
Color Dark blue
Composition Methanol, Methylene Blue
Storage Requirements <20°C
Stain Type Acid Fast Stain
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Methylene Blue
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash contaminated skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P370+P378-In case of fire: Use foam, carbon dioxide, dry powder or water fog to extinguish.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contains : methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.