missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue, pure, certified, residual water

Product Code. 10237273 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10237273 500 g Glass bottle 500g
10455081 25 g Glass bottle 25g
10175693 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10237273 Supplier Thermo Scientific Chemicals Supplier No. 414245000
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Methylene Blue, C16H24ClN3O3S, CAS Number-7220-79-3, Basic Blue 9, 25g, 1.21 to 1.7, Vapor Pressure: Negligible, 27, V,19, 259, CHEBI:6872, Green, 13 (Air - 1.0), 82% min., 200-515-2, 319.85, Pure, Warning, GHS H Statement: Harmful if swallowed. | CAS: 7220-79-3 | C16H24ClN3O3S | 373.90 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7220-79-3
Molecular Formula C16H24ClN3O3S
Molecular Weight (g/mol) 373.90
MDL Number MFCD00150008
InChI Key XQAXGZLFSSPBMK-UHFFFAOYSA-M
Synonym Basic Blue 9
PubChem CID 6099
ChEBI CHEBI:6872
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride
SMILES O.O.O.[Cl-].CN(C)C1=CC2=[S+]C3=CC(=CC=C3N=C2C=C1)N(C)C

Specifications

Melting Point 190.0°C
Absorption Ratio 1.21 to 1.7
Dye Content 82% min.
Infrared Spectrum Authentic
Lambda Max 663 to 667nm
Packaging Glass bottle
Vapor Pressure Vapor Pressure: Negligible
Beilstein 27, V,19, 259
Density 13 (Air = 1.0)
Merck Index 15, 6132
Color Green
Solubility Information Solubility in water: 40g/L (20°C). Other solubilities: in methanol: 430g/L
Quantity 500 g
Physical Form Fine Crystalline Powder
Formula Weight 319.85
Grade Pure
Chemical Name or Material Methylene Blue
Show More Show Less
compliance-icons
Product Identifier
  • Methylene Blue
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Methylene blue trihydrate, Methylene blue

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.