missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methylene Blue hydrate, 96+%, high purity biological stain

Product Code. p-29875 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
This item is not returnable. View return policy

Product Code. 10741891

Brand: Thermo Scientific Chemicals 229801000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16H18ClN3S
Molecular Weight (g/mol) 319.85
MDL Number MFCD00150006
InChI Key CXKWCBBOMKCUKX-UHFFFAOYSA-M
Synonym Basic Blue 9 hydrate, C.I. 52015 hydrate
PubChem CID 16211647
IUPAC Name [7-(dimethylamino)phenothiazin-3-ylidene]-dimethylazanium;chloride;hydrate
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C

Specifications

Melting Point 100.0°C to 110.0°C
Dye Content 96.0 to 103.0% (on dried substance)
Assay Percent Range 96+%
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol, ethanol
Infrared Spectrum Authentic
Formula Weight 319.85
Loss on Drying 8.0% to 22.0% (105°C)
Percent Purity ≥96%
Packaging Glass bottle
Color Green
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Methylene Blue hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Methylene Blue hydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Methylene Blue hydrate, Methylene blue

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.