missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Hydrate (>90%), TRC

Product Code. 30473814
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
30473814 100 g 100g
30464595 5 g 5g
30466554 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 30473814

Brand: Toronto Research Chemicals TRCM294245100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122965-43-9
Formula Weight 319.85
InChI Formula InChI=1S/C16H18N3S.ClH.H2O/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;;/h5-10H,1-4H3;1H;1H2/q+1;;/p-1
IUPAC Name 3-N,3-N,7-N,7-N-tetramethylphenothiazin-5-ium-3,7-diamine;chloride;hydrate
Molecular Formula C16 H18 Cl N3 S x H2 O
Molecular Weight (g/mol) 319.85
SMILES O.[Cl-].CN(C)c1ccc2nc3ccc(cc3[s+]c2c1)N(C)C
Synonym Methylene blue hydrate, 3,7-Bis(dimethylamino)phenothiazin-5-ium Chloride Hydrate
Chemical Name or Material Methylthioninium chloride hydrate

Specifications

Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.