missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Hydrate (>90%), TRC

Product Code. 30464595
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464595 5 g 5g
30473814 100 g 100g
30466554 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30464595 Supplier Toronto Research Chemicals Supplier No. TRCM2942455G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122965-43-9
Molecular Formula C16 H18 Cl N3 S x H2 O
Molecular Weight (g/mol) 319.85
Synonym Methylene blue hydrate, 3,7-Bis(dimethylamino)phenothiazin-5-ium Chloride Hydrate
IUPAC Name 3-N,3-N,7-N,7-N-tetramethylphenothiazin-5-ium-3,7-diamine;chloride;hydrate
SMILES O.[Cl-].CN(C)c1ccc2nc3ccc(cc3[s+]c2c1)N(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C16H18N3S.ClH.H2O/c1-18(2)11-5-7-13-15(9-11)20-16-10-12(19(3)4)6-8-14(16)17-13;;/h5-10H,1-4H3;1H;1H2/q+1;;/p-1
Formula Weight 319.85
Chemical Name or Material Methylthioninium chloride hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.