Learn More
Methyl methanesulfonate, 99%, Thermo Scientific Chemicals
£40.10 - £264.00
Chemical Identifiers
CAS | 66-27-3 |
---|---|
Molecular Formula | C2H6O3S |
Molecular Weight (g/mol) | 110.13 |
MDL Number | MFCD00007557 |
InChI Key | MBABOKRGFJTBAE-UHFFFAOYSA-N |
Synonym | methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate |
PubChem CID | 4156 |
ChEBI | CHEBI:25255 |
IUPAC Name | methyl methanesulfonate |
SMILES | COS(=O)(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10756571
|
Thermo Scientific Acros
156890050 |
5 g | Glass bottle |
£40.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10766571
|
Thermo Scientific Acros
156890250 |
25 g | Glass bottle |
£91.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696904
|
Thermo Scientific Acros
156891000 |
100 g | Glass bottle |
£264.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkyl transfer reagentChemical Identifiers
66-27-3 | |
110.13 | |
MBABOKRGFJTBAE-UHFFFAOYSA-N | |
4156 | |
methyl methanesulfonate |
C2H6O3S | |
MFCD00007557 | |
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate | |
CHEBI:25255 | |
COS(=O)(=O)C |
Specifications
66-27-3 | |
1.3000g/mL | |
104°C | |
98.5% min. (GC) | |
CH3SO3CH3 | |
MFCD00007557 | |
15, 6169 | |
methyl mesylate, methanesulfonic acid methyl ester, methanesulfonic acid, methyl ester, methylmethane sulfonate, methylmethansulfonat, methyl methanesulphonate, as-dimethyl sulfite, as-dimethyl sulphite, methylmethanesulfonate, methyl methansulfonate | |
MBABOKRGFJTBAE-UHFFFAOYSA-N | |
methyl methanesulfonate | |
4156 | |
110.13 | |
Liquid |
Colorless to Yellow | |
202.0°C to 203.0°C | |
Authentic | |
C2H6O3S | |
1.4130 to 1.4150 | |
04, 4 | |
1.3 | |
Solubility in water: 200g/L (20°C). Other solubilities: soluble in dmf | |
COS(=O)(=O)C | |
110.13 | |
CHEBI:25255 | |
99% | |
Methyl methanesulfonate |
Safety and Handling
GHS H Statement:
May cause cancer.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause damage to organs.
Suspected of causing genetic defects.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-625-
- Methyl methanesulfonate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H340-May cause genetic defects.
- H351-Suspected of causing cancer.
- H361-Suspected of damaging fertility or the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P201-Obtain special instructions before use.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only