missing translation for 'onlineSavingsMsg'
Learn More

Methyl malonyl chloride, 97%

Product Code. 10169921 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10169921 25 g Glass bottle 25g
10435872 5 g Glass bottle 5g
10413845 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10169921 Supplier Thermo Scientific Acros Supplier No. 414510250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37517-81-0 | C4H5ClO3 | 136.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-81-0
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.53
MDL Number MFCD00013657
InChI Key UTBCRHAMJFMIIR-UHFFFAOYSA-N
PubChem CID 123460
IUPAC Name methyl 3-chloro-3-oxopropanoate
SMILES COC(=O)CC(=O)Cl

Specifications

Color Yellow
Density 1.2730g/mL
Boiling Point 57.0°C to 59.0°C (12.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClCOCH2CO2CH3
Refractive Index 1.4310 to 1.4350
Quantity 25 g
Beilstein 02, IV, 1886
Specific Gravity 1.273
Synonym methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate
Formula Weight 136.53
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl malonyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl malonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.