missing translation for 'onlineSavingsMsg'
Learn More

Methyl cyclopropanecarboxylate, 98%

Product Code. 10005853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10005853 500 g 500g
10634792 25 g 25g
10091420 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10005853 Supplier Thermo Scientific Acros Supplier No. 126715000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2868-37-3 | C5H8O2 | 100.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2868-37-3
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001280
InChI Key PKAHQJNJPDVTDP-UHFFFAOYSA-N
Synonym methyl cyclopropane carboxylate, cyclopropanecarboxylic acid, methyl ester, methylcyclopropanecarboxylate, cyclopropanecarboxylic acid methyl ester, methyl cyclopropylcarboxylate, unii-5uq53f585u, mcpc, pubchem11835, ksc490a7l, 1-methylcyclopropanecarboxylate
PubChem CID 76122
IUPAC Name methyl cyclopropanecarboxylate
SMILES COC(=O)C1CC1

Specifications

Color Colorless
Density 0.9800g/mL
Boiling Point 119.0°C (764.0 mmHg)
Flash Point 17°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C3H5CO2CH3
Refractive Index 1.4170 to 1.4190
Quantity 500 g
Beilstein 09, I, 3
Specific Gravity 0.98
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols
Formula Weight 100.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl cyclopropanecarboxylate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl cyclopropanecarboxylate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.