missing translation for 'onlineSavingsMsg'
Learn More

Methyl chloroacetate, 98%, Thermo Scientific Chemicals

Catalog Number 10326600
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
96-34-4
C3H5ClO2
108.52
MFCD00000931
QABLOFMHHSOFRJ-UHFFFAOYSA-N
methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
7295
methyl 2-chloroacetate
COC(=O)CCl
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
96-34-4
C3H5ClO2
108.52
MFCD00000931
QABLOFMHHSOFRJ-UHFFFAOYSA-N
methyl chloroacetate, chloroacetic acid methyl ester, methyl monochloroacetate, methyl chloroethanoate, acetic acid, chloro-, methyl ester, methyl monochloracetate, methyl alpha-chloroacetate, methylchloroacetate, unii-450vsb182i, ccris 7749
7295
methyl 2-chloroacetate
COC(=O)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -33.0°C
Color Colorless
Density 1.2300g/mL
Boiling Point 130.0°C to 132.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CO2CH3
Refractive Index 1.4210 to 1.4230
Quantity 1 L
Beilstein 02,197
Merck Index 15,6113
Specific Gravity 1.23
Solubility Information Solubility in water: 28g/L (20°C). Other solubilities: soluble in methanol,ethanol,chloroform,,toluene,acetone,heptane,hexane,cyclohexane,,xylene and ether
Viscosity 1.23 mPa.s (20°C)
Formula Weight 108.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl chloroacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'