missing translation for 'onlineSavingsMsg'
Learn More

Methyle6-morpholinonicotinate, 97%, Thermo Scientific™

Product Code. 10294524
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10294524 5 g 5g
10427802 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10294524 Supplier Thermo Scientific Maybridge Supplier No. CC17522DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 132546-81-7
Molecular Formula C11H14N2O3
Molecular Weight (g/mol) 222.24
MDL Number MFCD03783347
InChI Key FFBPRHFKXVNLDV-UHFFFAOYSA-N
Synonym methyl 6-morpholinonicotinate, methyl 6-morpholin-4-yl pyridine-3-carboxylate, acmc-1by02, methyl 6-morpholin-4-ylnicotinate, methyl6-morpholinonicotinate, methyl 6-morpholin-4-yl-nicotinate, 6-4-morpholinyl-3-pyridinecarboxylic acid methyl ester
PubChem CID 2776466
IUPAC Name methyl 6-morpholin-4-ylpyridine-3-carboxylate
SMILES COC(=O)C1=CN=C(C=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 222.24
Percent Purity 97%
Chemical Name or Material Methyle6-morpholinonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.