missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-methoxy-2-indolecarboxylate, 98%, Thermo Scientific™

Product Code. 10612275
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 98081-83-5
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00134301
InChI Key OPUUCOLVBDQWEY-UHFFFAOYSA-N
Synonym methyl 6-methoxy-2-indolecarboxylate, 6-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl-6-methoxy-2-indolecarboxylate, methyl 6-methoxy-indole-2-carboxylate, 6-methoxyindole-2-carboxylic acid methyl ester, methyl 6-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester, methyl 6-methoxy-2-indolecarboxylat, pubchem7251, 6-methoxy-2-indolecarboxylate
PubChem CID 688173
IUPAC Name methyl 6-methoxy-1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=CC=C(OC)C=C2N1

Specifications

Melting Point 117°C to 119°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 205.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 6-methoxy-2-indolecarboxylate
compliance-icons
Product Identifier
  • Methyl 6-methoxy-2-indolecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'