missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-methoxy-2-indolecarboxylate, 98%, Thermo Scientific™

Product Code. 10255124 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255124 5 g Glass bottle 5g
10612275 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10255124 Supplier Thermo Scientific Acros Supplier No. 397920050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 98081-83-5
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD00134301
InChI Key OPUUCOLVBDQWEY-UHFFFAOYSA-N
Synonym methyl 6-methoxy-2-indolecarboxylate, 6-methoxy-1h-indole-2-carboxylic acid methyl ester, methyl-6-methoxy-2-indolecarboxylate, methyl 6-methoxy-indole-2-carboxylate, 6-methoxyindole-2-carboxylic acid methyl ester, methyl 6-methoxyindole-2-carboxylate, 1h-indole-2-carboxylic acid, 6-methoxy-, methyl ester, methyl 6-methoxy-2-indolecarboxylat, pubchem7251, 6-methoxy-2-indolecarboxylate
PubChem CID 688173
IUPAC Name methyl 6-methoxy-1H-indole-2-carboxylate
SMILES COC(=O)C1=CC2=CC=C(OC)C=C2N1

Specifications

Melting Point 117°C to 119°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 205.21
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 6-methoxy-2-indolecarboxylate
compliance-icons
Product Identifier
  • Methyl 6-methoxy-2-indolecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.