missing translation for 'onlineSavingsMsg'
Learn More

Methyle6-(diethylamino)nicotinate, 95%, Thermo Scientific™

Product Code. 10269542
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269542 1 g Amber Glass Bottle 1g
10622025 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10269542 Supplier Thermo Scientific Maybridge Supplier No. MO07007DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-37-2
Molecular Formula C11H16N2O2
Molecular Weight (g/mol) 208.261
MDL Number MFCD05664431
InChI Key MYJCIGZNMMQDRA-UHFFFAOYSA-N
Synonym methyl 6-diethylamino nicotinate, methyl 6-diethylamino pyridine-3-carboxylate
PubChem CID 2794746
IUPAC Name methyl 6-(diethylamino)pyridine-3-carboxylate
SMILES CCN(CC)C1=NC=C(C=C1)C(=O)OC

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.26
Percent Purity 95%
Chemical Name or Material Methyl 6-(diethylamino)nicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.