missing translation for 'onlineSavingsMsg'
Learn More

Methyl 6-amino-5-bromonicotinate, 98%, Thermo Scientific™

Product Code. 10351380
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 180340-70-9
Molecular Formula C7H8BrN2O2
Molecular Weight (g/mol) 232.06
MDL Number MFCD06659561
InChI Key LNTAAMVZKGYIIA-UHFFFAOYSA-O
Synonym methyl 6-amino-5-bromonicotinate, methyl 2-amino-3-bromo-5-pyridinecarboxylate, methyl 2-amino-3-bromopyridine-5-carboxylate, methyl-6-amino-5-bromonicotinate, 3-pyridinecarboxylic acid, 6-amino-5-bromo-, methyl ester, 6-amino-5-bromonicotinic acid methyl ester, pubchem9472, acmc-209egj, methyl 6-amino-5-bromo-pyridine-3-carboxylate, methyl-6-5-bromonicotinate
PubChem CID 2764099
IUPAC Name methyl 6-amino-5-bromopyridine-3-carboxylate
SMILES COC(=O)C1=C[NH+]=C(N)C(Br)=C1

Specifications

Melting Point 173.5°C to 175.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 231.05
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyl 6-amino-5-bromonicotinate
compliance-icons
Product Identifier
  • Methyl 6-amino-5-bromonicotinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'