missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-formyl-2-methyl-3-furoate, 97%, Thermo Scientific™

Product Code. 10089391
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10089391 1 g Amber Glass Bottle 1g
10495512 250 mg Amber Glass Bottle 250mg
10544984 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10089391 Supplier Thermo Scientific Maybridge Supplier No. MO00666DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 81661-26-9
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
MDL Number MFCD03407372
InChI Key WINMHOHYPKKTEM-UHFFFAOYSA-N
Synonym methyl 5-formyl-2-methyl-3-furoate, 3-furancarboxylicacid, 5-formyl-2-methyl-, methyl ester, methyl 5-formyl-2-methyl-3-furancarboxylate, 5-formyl-2-methyl-furan-3-carboxylic acid methyl ester, 3-furancarboxylic acid, 5-formyl-2-methyl-, methyl ester
PubChem CID 2779820
IUPAC Name methyl 5-formyl-2-methylfuran-3-carboxylate
SMILES CC1=C(C=C(O1)C=O)C(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 5-formyl-2-methyl-3-furoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.