missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-bromonicotinate, 97%, Maybridge™

Product Code. 10507684
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10507684

Brand: Thermo Scientific Maybridge RF04619DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 29681-44-5
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00173668
InChI Key AAJZXPWBILCHAW-UHFFFAOYSA-N
Synonym methyl 5-bromonicotinate, 5-bromonicotinic acid methyl ester, 5-bromo-3-pyridinecarboxylic acid methyl ester, 5-bromo nicotinic acid methyl ester, 3-pyridinecarboxylic acid, 5-bromo-, methyl ester, 3-bromo-5-methoxycarbonyl pyridine, 5-bromopyridine-3-carboxylic acid methyl ester, 5-bromo-nicotinic acid methyl ester, methyl 5-bromo-3-pyridinecarboxylate, methyl 3-bromo-5-pyridinecarboxylate
PubChem CID 699336
IUPAC Name methyl 5-bromopyridine-3-carboxylate
SMILES COC(=O)C1=CC(=CN=C1)Br

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 216.03
Percent Purity 97%
Chemical Name or Material Methyle5-bromonicotinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.