missing translation for 'onlineSavingsMsg'
Learn More

Methyle5-bromo-4-methyl-2-thiophenecarboxylate, 97%, Thermo Scientific™

Product Code. 10283022
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10283022 1 g Amber Glass Bottle 1g
10394332 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10283022 Supplier Thermo Scientific Maybridge Supplier No. CC29010DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 54796-47-3
Molecular Formula C7H7BrO2S
Molecular Weight (g/mol) 235.095
MDL Number MFCD04038959
InChI Key RJWZFHRMQRWEIH-UHFFFAOYSA-N
Synonym methyl 5-bromo-4-methyl-2-thiophenecarboxylate, methyl 5-bromo-4-methyl-2-thiophene carboxylate, acmc-209ljm, 5-bromo-2-methoxycarbonyl-4-methylthiophene, methyl 5-bromo-4-methyl-thiophene-2-carboxylate, 5-bromo-4-methyl-2-thiophenecarboxylic acid methyl ester, 2-thiophenecarboxylicacid, 5-bromo-4-methyl-, methyl ester, 5-bromo-4-methyl-thiophene-2-carboxylic acid methyl ester, 2-thiophenecarboxylic acid, 5-bromo-4-methyl-, methyl ester
PubChem CID 2795489
IUPAC Name methyl 5-bromo-4-methylthiophene-2-carboxylate
SMILES CC1=C(SC(=C1)C(=O)OC)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 235.1
Percent Purity 97%
Chemical Name or Material Methyl 5-bromo-4-methyl-2-thiophenecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.